Sodium 4-fluorophenylacetate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

PC450186-1G PC450186-5G
APOSPC450186-1GEA 227 EUR
APOSPC450186-1G APOSPC450186-5G
Sodium 4-fluorophenylacetate
Sodium 4-fluorophenylacetate
Formule: C₈H₆FO₂·Na
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD28054312
CAS nummer: 178666-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR