4-Fluoro-3-phenoxybenzaldehyde oxime

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

PC450194-5G PC450194-1G
APOSPC450194-5GEA 538 EUR
APOSPC450194-5G APOSPC450194-1G
4-Fluoro-3-phenoxybenzaldehyde oxime
4-Fluoro-3-phenoxybenzaldehyde oxime
Formule: C₁₃H₁₀FNO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD19442126
CAS nummer: 1192002-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR