2-(Bromomethyl)-6-fluorobenzonitrile 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

PC500549-10G PC500549-1G PC500549-5G
APOSPC500549-10GEA 0 EUR
APOSPC500549-10G APOSPC500549-1G APOSPC500549-5G
2-(Bromomethyl)-6-fluorobenzonitrile 97%
2-(Bromomethyl)-6-fluorobenzonitrile
Formule: C₈H₅BrFN
MW: 214.034 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD18394042
CAS nummer: 1261686-95-6
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR