5-Bromo-2,4-difluorostyrene 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
PC502004-1G
PC502004-250MG
PC502004-5G
APOSPC502004-1GEA
58.4
EUR
APOSPC502004-1G
APOSPC502004-250MG
APOSPC502004-5G
5-Bromo-2,4-difluorostyrene 95%
5-Bromo-2,4-difluorostyrene
Formule:
C₈H₅BrF₂ Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD29049257 CAS nummer: 1935371-62-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...