3-Bromo-5-fluorophenylisothiocyanate
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
PC503011-1G
PC503011-25G
PC503011-5G
PC503011-100G
APOSPC503011-1GEA
87.6
EUR
APOSPC503011-1G
APOSPC503011-25G
APOSPC503011-5G
APOSPC503011-100G
3-Bromo-5-fluorophenylisothiocyanate
3-Bromo-5-fluorophenylisothiocyanate
Formule:
C₇H₃BrFNS MW: 232.073 g/mol Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD32200432 CAS nummer: 1861560-52-2 UN: 2922 ADR: 8+6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...