2-Amino-4-nitrobenzotrifluoride 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PC5159-1G PC5159-250MG
APOSPC5159-1GEA 118 EUR
APOSPC5159-1G APOSPC5159-250MG
2-Amino-4-nitrobenzotrifluoride 97%
2-Amino-4-nitrobenzotrifluoride
Formule: C₇H₅F₃N₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03094261
CAS nummer: 393-49-7

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR