2-Amino-4-fluoroindole hydrochloride 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PC52232-1G PC52232-250MG PC52232-5G
APOSPC52232-1GEA 611 EUR
APOSPC52232-1G APOSPC52232-250MG APOSPC52232-5G
2-Amino-4-fluoroindole hydrochloride 95%
2-Amino-4-fluoroindole hydrochloride
Formule: C₈H₇FN₂·ClH
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD32638669
CAS nummer: 2514942-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR