1-Boc-4-(4-fluorophenyl)-4-carboxypiperidine 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PC530029-1G PC530029-250MG PC530029-500MG
APOSPC530029-1GEA 528 EUR
APOSPC530029-1G APOSPC530029-250MG APOSPC530029-500MG
1-Boc-4-(4-fluorophenyl)-4-carboxypiperidine 97%
1-Boc-4-(4-fluorophenyl)-4-carboxypiperidine
Formule: C₁₇H₂₂FNO₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD05861568
CAS nummer: 644981-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR