Zinc difluoromethanesulphinate
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC53016-1G PC53016-250MG PC53016-5G
APOSPC53016-1GEA 220 EUR
APOSPC53016-1G APOSPC53016-250MG APOSPC53016-5G
Zinc difluoromethanesulphinate
Zinc difluoromethanesulphinate
Formule: C₂H₂F₄O₄S₂Zn
MDL Nummer: MFCD23134675
CAS nummer: 1355729-38-2