3-Amino-6-(trifluoromethoxy)pyridine 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
6-(Trifluoromethoxy)pyridin-3-amine
PC53248-100MG
PC53248-1G
PC53248-250MG
APOSPC53248-100MGEA
0
EUR
APOSPC53248-100MG
APOSPC53248-1G
APOSPC53248-250MG
3-Amino-6-(trifluoromethoxy)pyridine 95%
3-Amino-6-(trifluoromethoxy)pyridine
Formule:
C₆H₅F₃N₂O Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD07374879 CAS nummer: 135900-33-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...