6-Bromo-4-fluoro-1H-indole

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

PC53423-1G PC53423-250MG
APOSPC53423-1GEA 267 EUR
APOSPC53423-1G APOSPC53423-250MG
6-Bromo-4-fluoro-1H-indole
6-Bromo-4-fluoro-1H-indole
Formule: C₈H₅BrFN
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD07781550
CAS nummer: 885520-59-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR