2,4-Dichloro-6-(4-fluorophenoxy)-1,3,5-triazine
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
PC7086-250MG
PC7086-100MG
APOSPC7086-250MGEA
0
EUR
APOSPC7086-250MG
APOSPC7086-100MG
2,4-Dichloro-6-(4-fluorophenoxy)-1,3,5-triazine
2,4-Dichloro-6-(4-fluorophenoxy)-1,3,5-triazine
Formule:
C₉H₄Cl₂FN₃O Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00117397 CAS nummer: 112748-46-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...