Trifluoroacetaldehyde monohydrate 72% in aqueous solution
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
PC7130-100G PC7130-10G PC7130-50G
APOSPC7130-100GEA 238 EUR
APOSPC7130-100G APOSPC7130-10G APOSPC7130-50G
Trifluoroacetaldehyde monohydrate 72% in aqueous solution
Trifluoroacetaldehyde monohydrate
A stable source of trifluoroacetaldehyde. Add the product dropwise to PPA @ 150-180C - distill off & react at once.
Formule: C₂HF₃O·H₂O
Kookpunt: 104…105 °C
Smeltpunt: 68…70 °C
Dichtheid: 1,468 g/cm³ (25 °C)
Vlampunt: –23 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00167359
CAS nummer: 33953-86-5
UN: 1989
ADR: 3,II