6-Fluoropyridine-2-carboxamide 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

PC7884-1G PC7884-250MG PC7884-5G
APOSPC7884-1GEA 39.5 EUR
APOSPC7884-1G APOSPC7884-250MG APOSPC7884-5G
6-Fluoropyridine-2-carboxamide 97%
6-Fluoropyridine-2-carboxamide
Formule: C₆H₅FN₂O
Smeltpunt: 134…137 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04038712
CAS nummer: 369-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR