4-Fluoro-2-isopropoxyaniline 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PC900142-1G PC900142-5G
APOSPC900142-1GEA 249 EUR
APOSPC900142-1G APOSPC900142-5G
4-Fluoro-2-isopropoxyaniline 98%
4-Fluoro-2-isopropoxyaniline
Formule: C₉H₁₂FNO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09738072
CAS nummer: 148583-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR