4-Ethoxy-3-fluorophenol 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC904078-1G
PC904078-250MG
APOSPC904078-1GEA
249
EUR
APOSPC904078-1G
APOSPC904078-250MG
4-Ethoxy-3-fluorophenol 95%
4-Ethoxy-3-fluorophenol
Formule:
C₈H₉FO₂ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD21984328 CAS nummer: 98121-48-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...