3-Bromo-5-fluoroisonicotinamide 97%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
PC904955-1G
PC904955-250MG
APOSPC904955-1GEA
176
EUR
APOSPC904955-1G
APOSPC904955-250MG
3-Bromo-5-fluoroisonicotinamide 97%
3-Bromo-5-fluoroisonicotinamide
Formule:
C₆H₄BrFN₂O Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD20524845 CAS nummer: 1353636-72-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...