4-(2-Fluorophenyl)phenol 97%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC909233-1G
PC909233-250MG
APOSPC909233-1GEA
541
EUR
APOSPC909233-1G
APOSPC909233-250MG
4-(2-Fluorophenyl)phenol 97%
4-(2-Fluorophenyl)phenol
Formule:
C₁₂H₉FO Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD06802292 CAS nummer: 321-62-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...