5-Fluoro-3,4-dihydro-2H-1,4-benzoxazine 98%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC910687-1G
PC910687-250MG
APOSPC910687-1GEA
811
EUR
APOSPC910687-1G
APOSPC910687-250MG
5-Fluoro-3,4-dihydro-2H-1,4-benzoxazine 98%
5-Fluoro-3,4-dihydro-2H-1,4-benzoxazine
Formule:
C₈H₈FNO Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD11878397 CAS nummer: 1067171-66-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...