α-Chloro-4-fluorobenzaldoxime
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
4-Fluoro-N-hydroxybenzimidoyl chloride
PC9299-10G
PC9299-50G
APOSPC9299-10GEA
435
EUR
APOSPC9299-10G
APOSPC9299-50G
α-Chloro-4-fluorobenzaldoxime
α-Chloro-4-fluorobenzaldoxime
Formule:
C₇H₅ClFNO MW: 173.572 g/mol Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD02179417 CAS nummer: 42202-95-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...