2-(2,6-Difluoro-3-propoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC99014-10G PC99014-1G PC99014-5G
APOSPC99014-10GEA 651 EUR
APOSPC99014-10G APOSPC99014-1G APOSPC99014-5G
2-(2,6-Difluoro-3-propoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-(2,6-Difluoro-3-propoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane