Eppendorf® Pipette Manager - Connected Pipette System
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1004000.001 4861000.970
613-6958EA 1110 EUR
613-6958 613-6959
Eppendorf® Pipette Manager - Connected Pipette System
Software Laboratoriumsoftware
Control Eppendorf's connected electronic pipettes via a dedicated WiFi for faster, more reproducible results. Remotely set up pipettes, receive pipetting technique guidance, digitally document activities, and more.
  • Connected electronic pipette system to remotely set up electronic pipettes, ease handling of challenging liquids and digitally document pipetting steps
  • Choose from predefined Eppendorf liquid types (e.g., ethanol or enzymes/antibodies) or create your own
  • Manage your pipette fleet more effectively and enhance collaboration with integrated features like Pipette set creation or liquid type Sharing
  • Gain additional pipetting modes for your Xplorer pipettes (Sequential Dispensing, Sequential Pipetting, Diluting, Reverse Pipetting, Multi Aspiration)
  • Easily update your Xplorer, Xplorer plus and Xplorer plus Move It pipette software with the Pipette Manager to never miss the latest features
All Eppendorf Xplorer™, Xplorer plus and Xplorer plus Move It™ electronic pipettes are compatible (when upgraded with the Eppendorf Xplorer connect WiFi module). Automatically document pipetting activities via Pipetting Records and easily export your recordings as comprehensive PDF document via USB.
The graphical user interface is based on the VisioNize™ touch interface concept that has been extensively tested by customers to reduce learning and adjustment time. The software-embedded Liquid Manager sets optimal pipetting settings (e.g., pre-wetting steps, speed) when working with viscous, volatile and other challenging liquid types.
Easily enter pipetting volumes and other parameters using the touch screen. Settings are immediately transferred to connected electronic pipettes, allowing you to start working right away.