Sampling scoops, PS, white, double-bagged, Sterileware®, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H36902-1010 H36904-1010 H36906-1010
231-0641EA 84.8 EUR
231-0641 231-0642 231-0643
Sampling scoops, PS, white, double-bagged, Sterileware®, SP Bel-Art
Scheppen
Ideal for multi-zone cleanrooms, these one-piece double-bagged Sterileware® sampling scoops are the perfect shape and size for taking samples, especially from wide mouth containers.
  • When placed on a flat surface they remain upright; can be used as weighing boats
  • 60 and 125 ml sampling scoops have a long handle that increases the distance between the hand and sample
  • FDA grade polystyrene safe for food, drug and cosmetic use
  • Gamma irradiated sterile to SAL 10⁻⁶
  • Lot stamped for accurate lot tracing, and shipped with a certificate of processing for
More Product Information
Order Now
Specificaties
Environmentally Preferable