L-Alanine
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
BIA0703-100G
BIA0703-1KG
BIA0703-500G
APOSBIA0703-100GEA
28.9
EUR
APOSBIA0703-100G
APOSBIA0703-1KG
APOSBIA0703-500G
L-Alanine
L-Alanine
USP, pyrogen free
Formule:
C₃H₇NO₂ Dichtheid: 1,432 g/cm³ (25 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00064410 CAS nummer: 56-41-7 EINECS: 200-273-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...