N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride, BAPNA, BANI, Bz-DL-Arg-pNA·HCl
BIB1056-1G BIB1056-5G
APOSBIB1056-1GEA 51.2 EUR
APOSBIB1056-1G APOSBIB1056-5G
N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride
N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride
Chromogenic substrate for trypsin and other proteolytic enzymes.
Formule: C₁₉H₂₃ClN₆O₄
MW: 434.881 g/mol
Smeltpunt: 272…276 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00012846
CAS nummer: 911-77-3