Oosporein
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
BIBR1071-1MG
BIBR1071-5MG
APOSBIBR1071-1MGEA
243
EUR
APOSBIBR1071-1MG
APOSBIBR1071-5MG
Oosporein
Oosporein
Formule:
C₁₄H₁₀O₈ Bewaar temperatuur: Vriezer |
MDL Nummer:
MFCD01570989 CAS nummer: 475-54-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...