Cefotetan

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

BIC0104-100MG BIC0104-250MG BIC0104-500MG
APOSBIC0104-100MGEA 22 EUR
APOSBIC0104-100MG APOSBIC0104-250MG APOSBIC0104-500MG
Cefotetan
Cefotetan
Formule: C₁₇H₁₇N₇O₈S₄
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00864983
CAS nummer: 69712-56-7
EINECS: 274-093-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR