Fumitremorgin C
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
BIF1010-10MG
BIF1010-1MG
BIF1010-5MG
APOSBIF1010-10MGEA
1910
EUR
APOSBIF1010-10MG
APOSBIF1010-1MG
APOSBIF1010-5MG
Fumitremorgin C
Fumitremorgin C
Formule:
C₂₂H₂₅N₃O₃ Bewaar temperatuur: Koelkast |
CAS nummer:
118974-02-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...