N-Octyl-β-D-glucopyranoside ≥99%, Ultrapure
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Octylglucopyranoside, OG, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triol, n-Octyl β-D-glucopyranoside, 1-O-n-Octyl-β-D-glucopyranoside, Octyl-β-D-glucopyranoside
BIMB1075-10G BIMB1075-1G BIMB1075-5G
APOSBIMB1075-10GEA 0 EUR
APOSBIMB1075-10G APOSBIMB1075-1G APOSBIMB1075-5G
N-Octyl-β-D-glucopyranoside ≥99%, Ultrapure
N-Octyl-β-D-glucopyranoside
Non-ionic detergent used for membrane protein solubisation. An advantage of OG is its low molecular weight which allows easy removal by dialysis.
Formule: C₁₄H₂₈O₆
MW: 292.37 g/mol
Smeltpunt: 103…104 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00063288
CAS nummer: 29836-26-8
EINECS: 249-887-8