Guanidiniumchloride, ultrapure
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride
BIMB3002-1KG BIMB3002-500G
APOSBIMB3002-1KGEA 176 EUR
APOSBIMB3002-1KG APOSBIMB3002-500G
Guanidiniumchloride, ultrapure
Guanidiniumchloride
  • Protein
Formule: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Smeltpunt: 185
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013026
CAS nummer: 50-01-1
Merck Index: 13,04578