(E/Z)-BIX02189

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 3-[[[3-[(Dimethylamino)methyl]phenyl]amino]phenylmethylene]-2,3-dihydro-N,N-dimethyl-2-oxo-1H-indole-6-carboxamide, BIX-02189 (Random Configuration), BIX-02189

BISN0224-1MG BISN0224-5MG BISN0224-10MG
APOSBISN0224-1MGEA 139 EUR
APOSBISN0224-1MG APOSBISN0224-5MG APOSBISN0224-10MG
(E/Z)-BIX02189
(E/Z)-BIX02189

MAPK/ERK Signalling Pathway

Formule: C₂₇H₂₈N₄O₂
MW: 440.54 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD18074528
CAS nummer: 1094614-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR