Kaempferol
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one
BISN0318-100MG
BISN0318-250MG
APOSBISN0318-100MGEA
110
EUR
APOSBISN0318-100MG
APOSBISN0318-250MG
Kaempferol
Kaempferol
Formule:
C₁₅H₁₀O₆ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00016938 CAS nummer: 520-18-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...