Dicopper selenide 99.99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

IN1582-25G IN1582-5G
APOSIN1582-25GEA 122 EUR
APOSIN1582-25G APOSIN1582-5G
Dicopper selenide 99.99%
Dicopper selenide
Formule: Cu₂Se
Smeltpunt: 1113 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00049908
CAS nummer: 20405-64-5
EINECS: 243-796-7
UN: 3283
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR