Erbium trichloride hexahydrate 99,9%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
IN1693-100G IN1693-25G
APOSIN1693-100GEA 351 EUR
APOSIN1693-100G APOSIN1693-25G
Erbium trichloride hexahydrate 99,9%
Erbium trichloride hexahydrate
Formule: Cl₃Er·6H₂O
MW: 381.71 g/mol
Smeltpunt: 774 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149693
CAS nummer: 10025-75-9
EINECS: 233-385-0