Palladium hydroxide on carbon 95% 20% Pd, wet support, powder
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
IN2807-1G IN2807-5G IN2807-25G
APOSIN2807-1GEA 0 EUR
APOSIN2807-1G APOSIN2807-5G APOSIN2807-25G
Palladium hydroxide on carbon 95% 20% Pd, wet support, powder
Palladium hydroxide
Formule: H₂O₂Pd
MW: 140.435 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064599
CAS nummer: 12135-22-7
EINECS: 235-219-2