1,2,3,4-Tetrahydroquinoxaline

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

OR01714-1G OR01714-5G OR01714-25G
APOSOR01714-1GEA 29.2 EUR
APOSOR01714-1G APOSOR01714-5G APOSOR01714-25G
1,2,3,4-Tetrahydroquinoxaline
1,2,3,4-Tetrahydroquinoxaline
Formule: C₈H₁₀N₂
MW: 134.18 g/mol
Smeltpunt: 92…98 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00047564
CAS nummer: 3476-89-9
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR