4-Isopropylthiophenol, Technical Grade
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
OR0781-100G
OR0781-25G
OR0781-500G
APOSOR0781-100GEA
58.4
EUR
APOSOR0781-100G
APOSOR0781-25G
APOSOR0781-500G
4-Isopropylthiophenol, Technical Grade
4-Isopropylthiophenol
Formule:
C₉H₁₂S MW: 152.26 g/mol Kookpunt: 99…100 °C/14mmHg Dichtheid: 0.979 g/cm³ (25 °C) Vlampunt: 106 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00041505 CAS nummer: 4946-14-9 UN: 2922 ADR: 8+6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...