m-Toluonitrile 98%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
m-Tolunitrile, m-Methylbenzonitrile, 3-Methylbenzonitrile
OR0828-25G
OR0828-1G
OR0828-2.5G
OR0828-5G
OR0828-10G
OR0828-50G
APOSOR0828-25GEA
42.4
EUR
APOSOR0828-25G
APOSOR0828-1G
APOSOR0828-2.5G
APOSOR0828-5G
APOSOR0828-10G
APOSOR0828-50G
m-Toluonitrile 98%
m-Toluonitrile
Formule:
C₈H₇N Kookpunt: 99…101 °C Smeltpunt: –23 °C Dichtheid: 0,987 g/cm³ (25 °C) Vlampunt: 86 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00001808 CAS nummer: 620-22-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...