Ethyl thiooxamate 97%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
2-Thiooxamic acid ethyl ester, Ethyl amino(thioxo)acetate, Ethyl 2-amino-2-thioxoacetate
OR10873-5G
OR10873-25G
OR10873-100G
APOSOR10873-5GEA
22
EUR
APOSOR10873-5G
APOSOR10873-25G
APOSOR10873-100G
Ethyl thiooxamate 97%
Ethyl thiooxamate
Formule:
C₄H₇NO₂S MW: 133.17 g/mol Smeltpunt: 62…65 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00074903 CAS nummer: 16982-21-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...