5-Bromopyridine-2-carbonitrile 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

OR11382-25G OR11382-5G
APOSOR11382-25GEA 49.7 EUR
APOSOR11382-25G APOSOR11382-5G
5-Bromopyridine-2-carbonitrile 97%
5-Bromopyridine-2-carbonitrile
Formule: C₆H₃BrN₂
Kookpunt: 100…110 °C/3mmHg
Smeltpunt: 124…132 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00234144
CAS nummer: 97483-77-7
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR