3-Methylthiopropylamine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 3-Aminopropyl Methyl Sulphide, 3-Methylmercaptopropylamine

OR12831-1G OR12831-5G
APOSOR12831-1GEA 108 EUR
APOSOR12831-1G APOSOR12831-5G
3-Methylthiopropylamine
3-Methylthiopropylamine
Formule: C₄H₁₁NS
MW: 105.20 g/mol
Kookpunt: 169 °C
Dichtheid: 0.938 g/cm³ (25 °C)
Vlampunt: 62 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00041898
CAS nummer: 4104-45-4
EINECS: 223-875-2
UN: 2735
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR