Cyclopentanesulphonyl chloride
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
OR12994-1G
OR12994-5G
OR12994-25G
APOSOR12994-1GEA
95
EUR
APOSOR12994-1G
APOSOR12994-5G
APOSOR12994-25G
Cyclopentanesulphonyl chloride
Cyclopentanesulphonyl chloride
Formule:
C₅H₉ClO₂S Dichtheid: 1,279 g/cm³ (25 °C) Vlampunt: >110 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD04117774 CAS nummer: 26394-17-2 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...