Hexamethylphosphoramide 98%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
OR13709-100G
OR13709-25G
APOSOR13709-100GEA
190
EUR
APOSOR13709-100G
APOSOR13709-25G
Hexamethylphosphoramide 98%
Hexamethylphosphoramide
Formule:
C₆H₁₈N₃OP Kookpunt: 230…232 °C/740mmHg Smeltpunt: 7 °C Dichtheid: 1,03 g/cm³ (25 °C) Vlampunt: 144 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00008303 CAS nummer: 680-31-9 EINECS: 211-653-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...