1-(4-Formylphenyl)pyrrolidine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

OR1386-1G OR1386-5G
APOSOR1386-1GEA 22 EUR
APOSOR1386-1G APOSOR1386-5G
1-(4-Formylphenyl)pyrrolidine
1-(4-Formylphenyl)pyrrolidine
Formule: C₁₁H₁₃NO
Smeltpunt: 81…85 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00015469
CAS nummer: 51980-54-2
EINECS: 257-570-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR