Methyl-3-bromo-4-hydroxy-5-nitrobenzenecarboxylate 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR14142-1G
OR14142-5G
APOSOR14142-1GEA
42.4
EUR
APOSOR14142-1G
APOSOR14142-5G
Methyl-3-bromo-4-hydroxy-5-nitrobenzenecarboxylate 95%
Methyl-3-bromo-4-hydroxy-5-nitrobenzenecarboxylate
Formule:
C₈H₆BrNO₅ Smeltpunt: 131…133 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD08689690 CAS nummer: 40258-72-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...