2-Butynoic acid ethyl ester

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

OR14510-1G OR14510-25G OR14510-5G
APOSOR14510-1GEA 0 EUR
APOSOR14510-1G APOSOR14510-25G APOSOR14510-5G
2-Butynoic acid ethyl ester
2-Butynoic acid ethyl ester
Formule: C₆H₈O₂
Kookpunt: 164 °C
Dichtheid: 0,97 g/cm³ (25 °C)
Vlampunt: 62 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00015182
CAS nummer: 4341-76-8
EINECS: 224-395-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR