2-(Piperazin-1-yl)benzaldehyde

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

OR1644-1G OR1644-5G
APOSOR1644-1GEA 0 EUR
APOSOR1644-1G APOSOR1644-5G
2-(Piperazin-1-yl)benzaldehyde
2-(Piperazin-1-yl)benzaldehyde
Formule: C₁₁H₁₄N₂O
Bewaar temperatuur: Kamertemperatuur
CAS nummer: 736991-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR