3,4,6-Trichloropyridazine
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR16890-5G
OR16890-25G
OR16890-100G
APOSOR16890-5GEA
22
EUR
APOSOR16890-5G
APOSOR16890-25G
APOSOR16890-100G
3,4,6-Trichloropyridazine
3,4,6-Trichloropyridazine
Formule:
C₄HCl₃N₂ Kookpunt: 96…97,5 °C/2mmHg Smeltpunt: 55…59 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00834957 CAS nummer: 6082-66-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...