4-Bromo-2-(bromomethyl)-1-iodobenzene 95+%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

OR17110-1G OR17110-250MG
APOSOR17110-1GEA 92.1 EUR
APOSOR17110-1G APOSOR17110-250MG
4-Bromo-2-(bromomethyl)-1-iodobenzene 95+%
4-Bromo-2-(bromomethyl)-1-iodobenzene
Formule: C₇H₅Br₂I
Kookpunt: 335 °C/760mmHg
Smeltpunt: 83…85 °C
Vlampunt: 156 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD12025360
CAS nummer: 495414-06-7
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR