Methyl-6-bromo-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-8-carboxylate 95+%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR18032-1G
OR18032-100MG
OR18032-250MG
APOSOR18032-1GEA
242
EUR
APOSOR18032-1G
APOSOR18032-100MG
APOSOR18032-250MG
Methyl-6-bromo-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-8-carboxylate 95+%
Methyl-6-bromo-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-8-carboxylate
Formule:
C₁₀H₈BrNO₄ Smeltpunt: 264…265 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD12196467 CAS nummer: 141761-85-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...